Bis(trimethylsilyl)methane ≥98%
Supplier: Ambeed
|
Danger
|
A607542-25G
A607542-10G
A607542-1G
A607542-5G
77572-978EA
121
USD
77572-978
77878-410
77878-411
77878-412
Bis(trimethylsilyl)methane ≥98%
Bis(trimethylsilyl)methane
Precaución: for Research Use Only
Formula:
C₇H₂₀Si₂ |
MDL Number:
MFCD00008275 CAS Number: 2117-28-4 UN: 1993 ADR: 3,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...