4-(1H-Pyrazol-4-yl)pyridine ≥95%

Supplier: Ambeed
Warning

A607029-1G A607029-250MG A607029-5G
77572-974EA 66 USD
77572-974 77749-251 77749-252
4-(1H-Pyrazol-4-yl)pyridine ≥95%
4-(1H-Pyrazol-4-yl)pyridine

Precaución: for Research Use Only

Formula: C₈H₇N₃
MDL Number: MFCD00480339
CAS Number: 19959-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR