2,3-Difluoro-5-nitrophenylboronic acid ≥98%

Supplier: Ambeed
Warning

A607002-1G A607002-250MG
77882-426EA 162 USD
77882-426 77882-427
2,3-Difluoro-5-nitrophenylboronic acid ≥98%
2,3-Difluoro-5-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BF₂NO₄
MDL Number: MFCD09027242
CAS Number: 957060-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR