tert-Butyl 8-amino-3-azabicyclo[3.2.1]octane-3-carboxylate ≥98%

Supplier: Ambeed
Warning

A606972-100MG A606972-1G
77673-644EA 57 USD
77673-644 77673-646
tert-Butyl 8-amino-3-azabicyclo[3.2.1]octane-3-carboxylate ≥98%
tert-Butyl 8-amino-3-azabicyclo[3.2.1]octane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MW: 226.31 g/mol
MDL Number: MFCD22566230
CAS Number: 1330763-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR