(E)-3-Chlorocinnamaldehyde ≥96%

Supplier: Ambeed
Warning

A606962-25G A606962-1G A606962-5G
77749-249EA 543 USD
77749-249 77749-248 77749-250
(E)-3-Chlorocinnamaldehyde ≥96%
(E)-3-Chlorocinnamaldehyde

Precaución: for Research Use Only

Formula: C₉H₇ClO
MW: 166.60 g/mol
MDL Number: MFCD07437882
CAS Number: 56578-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR