3-Chloro-4-fluoro-5-methylphenylboronic acid, pinacol ester ≥98%

Supplier: Ambeed
Warning

A598548-1G A598548-100MG A598548-250MG A598548-5G
77594-820EA 560.68 USD
77594-820 77613-862 77613-864 77686-518
3-Chloro-4-fluoro-5-methylphenylboronic acid, pinacol ester ≥98%
3-Chloro-4-fluoro-5-methylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BClFO₂
MDL Number: MFCD20231468
CAS Number: 1402238-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR