4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[c][1,2,5]thiadiazole ≥95%

Supplier: Ambeed
Warning

A598280-100MG A598280-250MG
77673-608EA 47 USD
77673-608 77673-610
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[c][1,2,5]thiadiazole ≥95%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[c][1,2,5]thiadiazole

Precaución: for Research Use Only

Formula: C₁₂H₁₅BN₂O₂S
MW: 262.13 g/mol
MDL Number: MFCD21604133
CAS Number: 1088118-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR