Dibutyl sulphide ≥98%

Supplier: Ambeed
Danger

Synonyms: Dibutylsulphide, Butyl sulphide, n-Butyl sulphide

A598007-25G A598007-500G
77572-926EA 18 USD
77572-926 77880-272
Dibutyl sulphide ≥98%
Dibutyl sulphide

Precaución: for Research Use Only

Formula: C₈H₁₈S
MW: 146.3 g/mol
Boiling Pt: 189
Melting Pt: -80
Flash Pt: 62
MDL Number: MFCD00009468
CAS Number: 544-40-1
UN: 3082
ADR: 9 Ⅲ
Merck Index: 12,01626

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR