2,2-Difluorocyclohexanamine hydrochloride ≥97%

Supplier: Ambeed
Warning

A597660-1G A597660-100MG A597660-250MG
77673-606EA 234.38 USD
77673-606 77884-440 77884-441
2,2-Difluorocyclohexanamine hydrochloride ≥97%
2,2-Difluorocyclohexanamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClF₂N
MDL Number: MFCD11878024
CAS Number: 921602-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR