2-Ethoxyethyl cyanoacetate ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxyethyl 2-cyanoacetate, Cyanoacetic Acid 2-Ethoxyethyl Ester

A597000-500G A597000-100G
77749-230EA 145 USD
77749-230 77749-229
2-Ethoxyethyl cyanoacetate ≥98%
2-Ethoxyethyl cyanoacetate

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃
MW: 157.17 g/mol
MDL Number: MFCD00071546
CAS Number: 32804-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR