1-(tert-Butoxycarbonyl)indoline-6-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A596223-100MG A596223-250MG
77684-465EA 67 USD
77684-465 77684-466
1-(tert-Butoxycarbonyl)indoline-6-carboxylic acid ≥95%
1-(tert-Butoxycarbonyl)indoline-6-carboxylic acid

Precaución: For research use only.

Formula: C₁₄H₁₇NO₄
MW: 263.28 g/mol
MDL Number: MFCD09878909
CAS Number: 208772-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR