4-(Difluoromethoxy)benzene-1,2-diamine ≥95%

Supplier: Ambeed
Warning

A595915-100MG A595915-1G
77893-201EA 80 USD
77893-201 77893-202
4-(Difluoromethoxy)benzene-1,2-diamine ≥95%
4-(Difluoromethoxy)benzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₇H₈F₂N₂O
MDL Number: MFCD15147144
CAS Number: 172282-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR