Methyl 2-(methylthio)benzo[d]oxazole-6-carboxylate ≥98%

Supplier: Ambeed
Warning

A595673-100MG A595673-1G A595673-250MG A595673-5G
77886-440EA 17 USD
77886-440 77886-441 77886-442 77886-443
Methyl 2-(methylthio)benzo[d]oxazole-6-carboxylate ≥98%
Methyl 2-(methylthio)benzo[d]oxazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃S
MDL Number: MFCD14706595
CAS Number: 1160490-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR