4-Amino-2-ethoxy-5-nitrobenzoic acid ≥98%

Supplier: Ambeed
Warning

A595521-100MG A595521-250MG
77893-199EA 10 USD
77893-199 77893-200
4-Amino-2-ethoxy-5-nitrobenzoic acid ≥98%
4-Amino-2-ethoxy-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₅
MDL Number: MFCD04114334
CAS Number: 86718-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR