Chaetocin 99+%

Supplier: Ambeed
Warning

Synonyms: (3S,3′S,5aR,5′aR,10bR,10′bR,11aS,11′aS)-2,2′,3,3′,5a,5′a,6,6′-Octahydro-3,3′-bis(hydroxymethyl)-2,2′-dimethyl[10b,10′b(11H,11′H)-bi-3,11a-epidithio-11aH-pyrazino[1′,2′:1,5]pyrrolo[2,3-b]indole]-1,1′,4,4′-tetrone

A595022-1MG A595022-10MG A595022-5MG
77617-504EA 44.29 USD
77617-504 77749-227 77749-228
Chaetocin 99+%
Chaetocin

Precaución: for Research Use Only

Formula: C₃₀H₂₈N₆O₆S₄
MW: 696.839 g/mol
MDL Number: MFCD00133163
CAS Number: 28097-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR