(S)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one hydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-5-Benzyl-2,2,3-trimethyl-4-imidazolidinone hydrochloride

A595003-1G A595003-250MG A595003-5G
77572-920EA 115 USD
77572-920 77673-600 77673-602
(S)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one hydrochloride ≥98%
(S)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₉ClN₂O
MW: 254.76 g/mol
MDL Number: MFCD03426983
CAS Number: 278173-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR