Bis(2-ethylhexyl)azelate ≥98%

Supplier: Ambeed
Warning

Synonyms: Dioctyl Azelate, Bis(2-ethylhexyl) nonanedioate, Azelaic acid di(2-ethylhexyl) ester, Azelaic acid dioctyl ester, Di(2-ethylhexyl) Azelate

A594617-5G A594617-100G
77572-916EA 21 USD
77572-916 77627-230
Bis(2-ethylhexyl)azelate ≥98%
Bis(2-ethylhexyl)azelate

Precaución: for Research Use Only

Formula: C₂₅H₄₈O₄
MDL Number: MFCD00040496
CAS Number: 103-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR