1,3-Dimethyl-2-(trifluoromethyl)benzene ≥95%

Supplier: Ambeed
Warning

A594358-100MG A594358-1G
77686-510EA 48 USD
77686-510 77686-511
1,3-Dimethyl-2-(trifluoromethyl)benzene ≥95%
1,3-Dimethyl-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₉F₃
MDL Number: MFCD14525527
CAS Number: 41818-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR