2-Phenyl-2-(pyrrolidin-1-yl)acetic acid ≥98%

Supplier: Ambeed
Warning

A585738-1G A585738-250MG
77669-546EA 65 USD
77669-546 77669-548
2-Phenyl-2-(pyrrolidin-1-yl)acetic acid ≥98%
2-Phenyl-2-(pyrrolidin-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₂
MW: 205.25 g/mol
MDL Number: MFCD05237231
CAS Number: 100390-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR