2-Bromo-3-iodoanisole ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1-iodo-3-methoxybenzene, 2-Bromo-3-iodophenyl methyl ether

A594310-1G A594310-5G
77684-461EA 88 USD
77684-461 77684-462
2-Bromo-3-iodoanisole ≥98%
2-Bromo-3-iodoanisole

Precaución: For research use only.

Formula: C₇H₆BrIO
MW: 312.93 g/mol
MDL Number: MFCD18391616
CAS Number: 74128-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR