Boc-6-Ahx-OSu ≥95%

Supplier: Ambeed
Warning

Synonyms: 6-[(tert-Butoxycarbonyl)amino]hexanoic acid N-Succinimidyl ester, Boc-ε-aminocaproic acid-OSu, 6-(Boc-amino)hexanoic acid N-Succinimidyl ester, Boc-NH-C6-NHS, Boc-6-Ahx-OSu, 6-[(tert-Butoxycarbonyl)amino]hexanoic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 6-(tert-butoxycarbonylamino)hexanoate

A594222-1G A594222-250MG
77572-914EA 22.66 USD
77572-914 77686-509
Boc-6-Ahx-OSu ≥95%
Boc-6-Ahx-OSu

Precaución: for Research Use Only

Formula: C₁₅H₂₄N₂O₆
MW: 328.365 g/mol
MDL Number: MFCD00237379
CAS Number: 51513-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR