2-(Piperidin-3-yl)ethan-1-ol hydrochloride ≥96%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Piperidyl)ethanol Hydrochloride

A594209-5G A594209-1G
77572-910EA 136.99 USD
77572-910 77673-594
2-(Piperidin-3-yl)ethan-1-ol hydrochloride ≥96%
2-(Piperidin-3-yl)ethan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅NO·ClH
MW: 165.661 g/mol
MDL Number: MFCD14583117
CAS Number: 16780-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR