5-(((1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl)oxy)-5-oxopentanoic acid ≥99% (by GC, mixture of isomers)

Supplier: Ambeed
Warning

Synonyms: (L)-Monomenthane-3-yl carbonate

A585552-100G A585552-25G A585552-5G
77613-810EA 206 USD
77613-810 77613-812 77684-444
5-(((1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl)oxy)-5-oxopentanoic acid ≥99% (by GC, mixture of isomers)
5-(((1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl)oxy)-5-oxopentanoic acid

Precaución: For research use only.

Formula: C₁₅H₂₆O₄
MW: 270.36 g/mol
MDL Number: MFCD13194764
CAS Number: 220621-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR