2-(Piperazin-1-yl)acetonitrile dihydrochloride ≥98%

Supplier: Ambeed
Warning

A594181-1G A594181-5G
77622-382EA 47.38 USD
77622-382 77783-488
2-(Piperazin-1-yl)acetonitrile dihydrochloride ≥98%
2-(Piperazin-1-yl)acetonitrile dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃Cl₂N₃
MDL Number: MFCD16547616
CAS Number: 1044707-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR