3-Pyrroline-2-one ≥90% (mixture of isomers)
Supplier: Ambeed
|
Warning
|
A594152-250MG
A594152-100MG
A594152-1G
A594152-5G
77572-908EA
99
USD
77572-908
77893-189
77893-190
77893-191
3-Pyrroline-2-one ≥90% (mixture of isomers)
3-Pyrroline-2-one
Precaución: for Research Use Only
Formula:
C₄H₅NO |
MDL Number:
MFCD06658513 CAS Number: 4031-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...