Potassium(tert-butylaminomethyl)trifluoroborate ≥98%

Supplier: Ambeed
Danger

Synonyms: Potassium ((tert-butylamino)methyl)trifluoroborate

A593857-1G A593857-250MG A593857-5G
77893-186EA 218 USD
77893-186 77893-187 77893-188
Potassium(tert-butylaminomethyl)trifluoroborate ≥98%
Potassium (tert-butylaminomethyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₅H₁₂BF₃KN
MW: 193.600 g/mol
MDL Number: MFCD10566510
CAS Number: 1256366-05-8
UN: 1759
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR