3,5-Difluoro-2-methoxyaniline ≥96%, Ambeed.Inc

Supplier: Ambeed
Danger

A585122-5G A585122-1G
77684-443EA 116 USD
77684-443 77893-175
3,5-Difluoro-2-methoxyaniline ≥96%, Ambeed.Inc
3,5-Difluoro-2-methoxyaniline

Precaución: For research use only.

Formula: C₇H₇F₂NO
MDL Number: MFCD09832386
CAS Number: 41860-67-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR