1,5-Hexadiene-3,4-diol ≥95% stabilized

Supplier: Ambeed
Danger

Synonyms: 3,4-Dihydroxy-1,5-hexadiene, 1,2-Divinylglycol, Divinylethylene Glycol

Stabilization: Stabilised with hydroquinone (HQ)

A593279-1G A593279-25G
77572-900EA 7.21 USD
77572-900 77572-902
1,5-Hexadiene-3,4-diol ≥95% stabilized
1,5-Hexadiene-3,4-diol

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD00081129
CAS Number: 1069-23-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR