Ethyl 7-bromoquinoline-3-carboxylate ≥98%

Supplier: Ambeed
Warning

A593220-250MG A593220-1G
77613-844EA 20.6 USD
77613-844 77684-460
Ethyl 7-bromoquinoline-3-carboxylate ≥98%
Ethyl 7-bromoquinoline-3-carboxylate

Precaución: For research use only.

Formula: C₁₂H₁₀BrNO₂
MW: 280.12 g/mol
MDL Number: MFCD16657753
CAS Number: 1226762-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR