2-(((3,5-Dimethylpyridin-2-yl)methyl)sulfinyl)-5-methoxy-1H-benzo[d]imidazole ≥95%

Supplier: Ambeed
Warning

A592645-10MG A592645-1MG A592645-5MG
77613-836EA 238.96 USD
77613-836 77613-838 77613-840
2-(((3,5-Dimethylpyridin-2-yl)methyl)sulfinyl)-5-methoxy-1H-benzo[d]imidazole ≥95%
2-(((3,5-Dimethylpyridin-2-yl)methyl)sulfinyl)-5-methoxy-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₆H₁₇N₃O₂S
MDL Number: MFCD08272115
CAS Number: 110374-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR