(5-Cyanobenzo[b]thiophen-2-yl)boronic acid 95+%

Supplier: Ambeed
Warning

A592067-100MG A592067-1G A592067-250MG A592067-50MG
77891-173EA 60 USD
77891-173 77891-174 77891-175 77891-176
(5-Cyanobenzo[b]thiophen-2-yl)boronic acid 95+%
(5-Cyanobenzo[b]thiophen-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₆BNO₂S
MDL Number: MFCD15143421
CAS Number: 1142946-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR