Pyrrolo[1,2-c]pyrimidine-3-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A583725-1G A583725-100MG A583725-5G
77669-536EA 111 USD
77669-536 77872-960 77872-961
Pyrrolo[1,2-c]pyrimidine-3-carboxylic acid ≥98%
Pyrrolo[1,2-c]pyrimidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD09878579
CAS Number: 251102-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR