Prunin ≥98%

Supplier: Ambeed
Danger

Synonyms: (S)-5-Hydroxy-2-(4-hydroxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one

A591934-10MG A591934-1MG A591934-5MG
77602-472EA 285.98 USD
77602-472 77602-474 77602-476
Prunin ≥98%
Prunin

Precaución: for Research Use Only

Formula: C₂₁H₂₂O₁₀
MDL Number: MFCD00210548
CAS Number: 529-55-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR