o-Cresolphthalein complexone indicator for complexometry

Supplier: Ambeed
Warning

A591881-5G A591881-25G
77602-470EA 27.68 USD
77602-470 77882-400
o-Cresolphthalein complexone indicator for complexometry
o-Cresolphthalein complexone

Precaución: for Research Use Only

Formula: C₃₂H₃₂N₂O₁₂
MDL Number: MFCD00005911
CAS Number: 2411-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR