3-(p-Tolyl)acrylaldehyde ≥97%

Supplier: Ambeed
Warning

A582979-25G A582979-5G
77749-199EA 117 USD
77749-199 77749-200
3-(p-Tolyl)acrylaldehyde ≥97%
3-(p-Tolyl)acrylaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀O
MW: 146.1858 g/mol
CAS Number: 1504-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR