tert-Butyl ((5-(aminomethyl)pyridin-2-yl)methyl)carbamate ≥95%

Supplier: Ambeed
Warning

A582945-100MG A582945-250MG
77602-446EA 348.5 USD
77602-446 77669-532
tert-Butyl ((5-(aminomethyl)pyridin-2-yl)methyl)carbamate ≥95%
tert-Butyl ((5-(aminomethyl)pyridin-2-yl)methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₉N₃O₂
MDL Number: MFCD28368809
CAS Number: 654679-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR