3,4-Dimethyl-2-hydroxybenzoic acid ≥97%

Supplier: Ambeed
Warning

A590335-1G A590335-250MG A590335-5G
77594-796EA 237.8 USD
77594-796 77747-227 77747-228
3,4-Dimethyl-2-hydroxybenzoic acid ≥97%
3,4-Dimethyl-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD11203625
CAS Number: 58138-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR