Fmoc-3,4-dehydroPro-OH ≥97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-2,5-dihydro-1H-pyrrole-2-carboxylic acid, Fmoc-3,4-dehydro-Pro-OH

A590224-100MG A590224-250MG A590224-5G
77880-261EA 79 USD
77880-261 77880-262 77880-263
Fmoc-3,4-dehydroPro-OH ≥97%
Fmoc-3,4-dehydroPro-OH

Precaución: for Research Use Only

Formula: C₂₀H₁₇NO₄
MW: 335.359 g/mol
MDL Number: MFCD00151940
CAS Number: 135837-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR