2,3,5-Tri-O-benzyl-D-ribofuranose ≥95%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R)-2,3,5-Tris(benzyloxy)-4-hydroxypentanal

A590172-5G A590172-1G
77617-482EA 53.56 USD
77617-482 77747-226
2,3,5-Tri-O-benzyl-D-ribofuranose ≥95%
2,3,5-Tri-O-benzyl-D-ribofuranose

Precaución: for Research Use Only

Formula: C₂₆H₂₈O₅
MW: 420.50 g/mol
MDL Number: MFCD03425640
CAS Number: 54623-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR