2-Hydroxy-8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene ≥97%

Supplier: Ambeed
Warning

A582070-1G A582070-250MG
77617-454EA 509.85 USD
77617-454 77747-207
2-Hydroxy-8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene ≥97%
2-Hydroxy-8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene

Precaución: for Research Use Only

Formula: C₁₆H₁₉BO₃
MDL Number: MFCD12827861
CAS Number: 1260151-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR