2-Methyl-[1,2,4]triazolo[1,5-a]pyridin-8-amine ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A581779-5G A581779-1G A581779-250MG
77684-437EA 479 USD
77684-437 77891-155 77891-156
2-Methyl-[1,2,4]triazolo[1,5-a]pyridin-8-amine ≥95%, Ambeed.Inc
2-Methyl-[1,2,4]triazolo[1,5-a]pyridin-8-amine

Precaución: For research use only.

Formula: C₇H₈N₄
MDL Number: MFCD16547421
CAS Number: 7169-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR