1-Chloro-2,5-difluoro-4-nitrobenzene ≥98%

Supplier: Ambeed
Warning

A589253-5G A589253-10G A589253-1G
77594-792EA 39.98 USD
77594-792 77602-458 77872-970
1-Chloro-2,5-difluoro-4-nitrobenzene ≥98%
1-Chloro-2,5-difluoro-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClF₂NO₂
MDL Number: MFCD08276209
CAS Number: 578-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR