Tetraacid ≥97%

Supplier: Ambeed
Warning

Synonyms: 1,3-bis(carboxyethoxy)-2,2-bis(carboxyethoxy)propane, 3,3'-((2,2-Bis((2-carboxyethoxy)methyl)propane-1,3-diyl)bis(oxy))dipropanoic acid, 3-[3-(2-carboxyethoxy)-2,2-bis(2-carboxyethoxymethyl)propoxy]propanoic acid

A589196-250MG A589196-1G
77598-588EA 75.85 USD
77598-588 77622-378
Tetraacid ≥97%
Tetraacid

Precaución: for Research Use Only

Formula: C₁₇H₂₈O₁₂
MW: 424.397 g/mol
MDL Number: MFCD08669605
CAS Number: 35638-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR