2-Hydroxy-2,3-propanetricarboxylic acid, potassium salt(1:x) ≥99%

Supplier: Ambeed
Warning

A570240-500G A570240-100G
77669-506EA 12 USD
77669-506 77880-228
2-Hydroxy-2,3-propanetricarboxylic acid, potassium salt(1:x) ≥99%
2-Hydroxy-2,3-propanetricarboxylic acid, potassium salt(1:x)

Precaución: for Research Use Only

Formula: C₆H₅K₃O₇
MW: 306.39 g/mol
MDL Number: MFCD00036362
CAS Number: 7778-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR