Sodium (2S,4R,8S,E)-8-((2S,2'R,4'S,5R,5'S,7R,7'R,8R,9S,9'S,10'R)-9-hydroxy-2'-((2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyltetrahydro-2H-pyran-2-yl)-9'-(((2S,5S,6R)-5-methoxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2,4',8,10'-tetramethyl-1,1',6,6'-tetraoxa[2,7'-bispiro[4.5]decan]-7-yl)-2,4,6-trimethyl-5-oxonon-6-enoate ≥98%
Supplier: Ambeed
|
Danger
|
A581200-10MG
A581200-5MG
77617-450EA
297.67
USD
77617-450
77617-452
Sodium (2S,4R,8S,E)-8-((2S,2'R,4'S,5R,5'S,7R,7'R,8R,9S,9'S,10'R)-9-hydroxy-2'-((2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyltetrahydro-2H-pyran-2-yl)-9'-(((2S,5S,6R)-5-methoxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2,4',8,10'-tetramethyl-1,1',6,6'-tetraoxa[2,7'-bispiro[4.5]decan]-7-yl)-2,4,6-trimethyl-5-oxonon-6-enoate ≥98%
Sodium (2S,4R,8S,E)-8-((2S,2'R,4'S,5R,5'S,7R,7'R,8R,9S,9'S,10'R)-9-hydroxy-2'-((2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyltetrahydro-2H-pyran-2-yl)-9'-(((2S,5S,6R)-5-methoxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2,4',8,10'-tetramethyl-1,1',6,6
Precaución: for Research Use Only
Formula:
C₄₇H₇₇NaO₁₄ MW: 889.1 g/mol |
MDL Number:
MFCD02874368 CAS Number: 65101-87-3 UN: 2811 ADR: 6.1,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...