4-Amino-3-fluoro-2-methylphenol ≥95%

Supplier: Ambeed
Warning

A581182-5G A581182-1G
77749-198EA 656 USD
77749-198 77749-197
4-Amino-3-fluoro-2-methylphenol ≥95%
4-Amino-3-fluoro-2-methylphenol

Precaución: for Research Use Only

Formula: C₇H₈FNO
MW: 141.14 g/mol
MDL Number: MFCD19207913
CAS Number: 1020173-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR