Cefaclor ≥98%

Supplier: Ambeed
Danger

Synonyms: (6R,7R)-7-((R)-2-Amino-2-phenylacetamido)-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

A580945-5G A580945-1G A580945-25G A580945-500G A580945-100G A580945-250MG
77594-766EA 49.2 USD
77594-766 77602-442 77602-444 77627-200 77627-196 77627-198
Cefaclor ≥98%
Cefaclor

Precaución: for Research Use Only

Formula: C₁₅H₁₄ClN₃O₄S
MW: 385.8 g/mol
MDL Number: MFCD00151471
CAS Number: 53994-73-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR