3-(Pyrrolidin-3-yl)pyridine dihydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-(3-Pyrrolidinyl)pyridine dihydrochloride

A569801-100MG A569801-1G
77673-522EA 73 USD
77673-522 77673-524
3-(Pyrrolidin-3-yl)pyridine dihydrochloride ≥98%
3-(Pyrrolidin-3-yl)pyridine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄Cl₂N₂
MW: 221.127 g/mol
MDL Number: MFCD02089474
CAS Number: 635309-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR