5-Amino-4-bromo-1-methyl-1H-pyrazole ≥97%

Supplier: Ambeed
Warning

A580725-1G A580725-5G
77884-414EA 32 USD
77884-414 77884-415
5-Amino-4-bromo-1-methyl-1H-pyrazole ≥97%
5-Amino-4-bromo-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₆BrN₃
MDL Number: MFCD00068157
CAS Number: 105675-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR