3-Chloro-5-fluorosalicylaldehyde ≥97%

Supplier: Ambeed
Danger

A569600-1G A569600-25G
77613-778EA 24.72 USD
77613-778 77673-520
3-Chloro-5-fluorosalicylaldehyde ≥97%
3-Chloro-5-fluorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClFO₂
MDL Number: MFCD04039291
CAS Number: 82128-69-6
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR